Learn More
Aviva Systems Biology Corp MRGPRX4 Antibody - C-terminal region (ARP59927_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP59927_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against MRGPRX4. It was validated on Western Blot. . Alternate names: GPCR, MGC129753, MGC129754, MRGX4, SNSR6ApplicationWB. Species reactivity: Human, Mouse, Rabbit
Catalog No. 50-286-880