Learn More
Medchemexpress LLC Mrgprx4 modulator-1 | 2492596-61-7 | 98.1% | C16H13ClF3NO3 | 10MG
SDP
Catalog No. 5000259426
Encompass_Preferred
MRGPRX4 modulator-1 is a small-molecule modulator of the Mas-related G-protein coupled receptor X4 used as a research tool for in vitro pharmacology and receptor signaling studies. It is supplied as a solid and as ready-to-use DMSO solutions, and is characterized by defined chemical identity, high purity, and documented storage recommendations.
  • Modulates MRGPRX4 receptor activity in vitro.
  • Available as solid and DMSO solution formats for flexible use.
  • High purity suitable for research applications.
  • Documented chemical identity (CAS 2492596-61-7) and formula C16H13ClF3NO3.
  • Stable when stored under recommended conditions for extended shelf life.
Catalog No. 50-002-59426 Supplier Medchemexpress LLC Supplier No. HY14599710MG
MRGPRX4 modulator-1 is a small-molecule modulator of the Mas-related G-protein coupled receptor X4 used as a research tool for in vitro pharmacology and receptor signaling studies. It is supplied as a solid and as ready-to-use DMSO solutions, and is characterized by defined chemical identity, high purity, and documented storage recommendations.
  • Modulates MRGPRX4 receptor activity in vitro.
  • Available as solid and DMSO solution formats for flexible use.
  • High purity suitable for research applications.
  • Documented chemical identity (CAS 2492596-61-7) and formula C16H13ClF3NO3.
  • Stable when stored under recommended conditions for extended shelf life.