Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC MRGPRX4 modulator-1 (compound 31-2) | 2492596-61-7 | 98.1% | 359.73 g·mol⁻¹ | C16H13ClF3NO3 | 5 MG
SDP

Catalog No. 5000223202
Encompass_Preferred

MRGPRX4 modulator-1 is a small-molecule antagonist of the Mas-related G-protein coupled receptor X4 (MRGPRX4) intended for in vitro and preclinical research into itch, pain, and immune-related pathways. The compound is supplied as a solid and as a DMSO solution and is characterized by potency suitable for biochemical and cellular assays.

  • Antagonist activity with IC50 <100 nM for MRGPRX4.
  • Molecular formula C16H13ClF3NO3 and molecular weight 359.73 g·mol⁻¹.
  • High purity around 98% as reported in documentation.
  • Available as solid milligram quantities and as a 10 mM solution in DMSO.
  • Appearance white to light yellow solid.
  • Storage: powder at -20°C (long term) or 4°C (short term); in solvent at -80°C or -20°C for shorter periods.
  • Intended for biochemical and cellular research into itch, pain, and immune pathways.

Catalog No. 50-002-23202 Supplier Medchemexpress LLC Supplier No. HY1459975MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MRGPRX4 modulator-1 is a small-molecule antagonist of the Mas-related G-protein coupled receptor X4 (MRGPRX4) intended for in vitro and preclinical research into itch, pain, and immune-related pathways. The compound is supplied as a solid and as a DMSO solution and is characterized by potency suitable for biochemical and cellular assays.

  • Antagonist activity with IC50 <100 nM for MRGPRX4.
  • Molecular formula C16H13ClF3NO3 and molecular weight 359.73 g·mol⁻¹.
  • High purity around 98% as reported in documentation.
  • Available as solid milligram quantities and as a 10 mM solution in DMSO.
  • Appearance white to light yellow solid.
  • Storage: powder at -20°C (long term) or 4°C (short term); in solvent at -80°C or -20°C for shorter periods.
  • Intended for biochemical and cellular research into itch, pain, and immune pathways.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.