Learn More
Electron Microscopy Sciences MRI Cayman Lab Island 4 Foot Lab Cart
SDP
Catalog No. 501927279
Encompass_Preferred
White Polyethylene MRI Cayman Lab Island 4 Foot Lab Cart is constructed from nonferrous, durable, resistant, high density polyethylene with a large amount of storage and work space. The integrated locking mechanism can be locked with the use of a padlock (not included) that keeps the tambour door securely locked in place. Inside the MRI Cayman Lab Island there is a generous amount of storage space with a built-in shelf. Even fully loaded, the MRI Cayman Lab Island can be easily maneuvered by one person with the aid of its 5 inch casters. Cleans with soap and water, bleach or mild solvents. Perfect for MRI areas and other situations MR Conditional certifications are required.
Catalog No. 50-192-7279 Supplier Electron Microscopy Sciences Supplier No. 6438641
White Polyethylene MRI Cayman Lab Island 4 Foot Lab Cart is constructed from nonferrous, durable, resistant, high density polyethylene with a large amount of storage and work space. The integrated locking mechanism can be locked with the use of a padlock (not included) that keeps the tambour door securely locked in place. Inside the MRI Cayman Lab Island there is a generous amount of storage space with a built-in shelf. Even fully loaded, the MRI Cayman Lab Island can be easily maneuvered by one person with the aid of its 5 inch casters. Cleans with soap and water, bleach or mild solvents. Perfect for MRI areas and other situations MR Conditional certifications are required.