Learn More
TrippNT MRI Lab Storage Shelf with 5 Bins
SDP
Supplier:  TrippNT 51888
Encompass_Preferred
TrippNT's MRI Lab Storage Shelf with 5 Bins presents a clean image and orderly storage of MRI supplies. Promotes safety by reducing clutter. 5 Compartments store high quantities of small consumables and the top shelf adds flexible storage for bottles, gloves, wipers or chemicals. PVC construction is MRI Safe.
Catalog No. NC0809277