Learn More
TrippNT MRI EXTRA WIDE CORE CART
SDP
Supplier:  TrippNT 51731
Encompass_Preferred
TrippNT's MRI Extra Wide Core Cart offers secures, flexible, and mobile storage that is MR safe. This cart has an extra wide flat work surface with built-in handles for easy maneuverability. Non-ferrous, white polyethylene construction with bulk storage and 3 large drawers. Ships fully assembled.
Catalog No. NC3590023