Learn More
Electron Microscopy Sciences MRI Maui Lab Island 2 Foot Lab Cart
SDP
Catalog No. 501927278
Encompass_Preferred
White Polyethylene MRI Maui Lab Island 2 Foot Lab Cart offers flexible storage and is designed for those who need a large amount of storage and workspace many carts don't offer. It is constructed from white, nonferrous, durable, resistant, high density polyethylene and and can be cleaned with soap and water, bleach, or mild solvents. The integrated locking mechanism can be locked with the use of a padlock (not included) that keeps the tambour door securely locked in place until the lock is disengaged allowing the door to roll up and into the cart. Inside the MRI Cayman features a generous amount of storage space with a built-in shelf that lets you keep the supplies that you use most closest to your reach. Cart features 5 inch non marking casters with two locking. Even fully loaded, the MRI Cayman can be easily maneuvered by one person with the aid of its 5 inch casters.
Catalog No. 50-192-7278 Supplier Electron Microscopy Sciences Supplier No. 6438640
White Polyethylene MRI Maui Lab Island 2 Foot Lab Cart offers flexible storage and is designed for those who need a large amount of storage and workspace many carts don't offer. It is constructed from white, nonferrous, durable, resistant, high density polyethylene and and can be cleaned with soap and water, bleach, or mild solvents. The integrated locking mechanism can be locked with the use of a padlock (not included) that keeps the tambour door securely locked in place until the lock is disengaged allowing the door to roll up and into the cart. Inside the MRI Cayman features a generous amount of storage space with a built-in shelf that lets you keep the supplies that you use most closest to your reach. Cart features 5 inch non marking casters with two locking. Even fully loaded, the MRI Cayman can be easily maneuvered by one person with the aid of its 5 inch casters.