Learn More
World Precision Instrument MRI Microflex Breather
SDP
Catalog No. 502538858
Encompass
Designed for use with MRI, x-ray and other imaging equipment. Low profile, composite plastic construction, feature vertical and angular adjustments for precise positioning.
Catalog No. 50-253-8858 Supplier World Precision Instrument Supplier No. EZ103MX
Designed for use with MRI, x-ray and other imaging equipment. Low profile, composite plastic construction, feature vertical and angular adjustments for precise positioning.