Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Marketlab MRI Multipurpose Cart

Catalog No. 22505121
Change view
Click to view available options
Dimensions (D x W x H):
23.75 x 15.75 x 34.62 in.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (D x W x H)
22-505-121 23.75 x 15.75 x 34.62 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 22-505-121 Supplier Marketlab Supplier No. 39993
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MR-conditional to 3T and designed for storing frequently used supplies nearby.

  • Multipurpose cart is MR-conditional to 3T
  • Nonferrous hardware and components
  • Use the included trays or stock with your own
  • Five shelves
  • Fixed, predetermined height-designated integral side rails
  • White EPVC Sintra™
  • Flat top work surface
  • Four 2 in. casters, two locking

Specifications

Product Type Phlebotomy Cart
Dimensions (D x W x H) 23.75 x 15.75 x 34.62 in.
Material PVC
No. of Shelves 4
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.