Learn More
Marketlab MRI Safe Slimline Headphones, Corded
Catalog No. 22505347
Change view
Click to view available options
Quantity:
1 Ea.
Catalog No. Quantity
22-505-347 1 Ea.
1 options
Catalog No. 22-505-347 Supplier Marketlab Supplier No. 139145
Add to Cart
Add to Cart
Help reduce patient anxiety by letting them focus on an audiobook or music during an MRI instead of the loud machine sounds.
  • Fit most head coils and adjust to fit most patients with its quick-click adjustable headband
  • When paired with earplugs, they effectively reduce noise by 29 dB
  • Must be used in conjunction with earplugs that have an NRR rating of 29 or more to meet OSHA standards
  • 29 decibels (29 dB NRR) of noise reduction when used with earplugs
  • Non-magnetic
  • Non-deforming, quick-click adjustable headband fits most patients
  • Ventilated inner headband
  • Thick foam padding enhances patient comfort
  • 1-year manufacturer warranty
Specifications
Product Type Over the Ear Headset
Quantity 1 Ea.