Learn More
Medchemexpress LLC MRIA9 | 2750707-05-0 | 98.9% | 496.92 | 50 MG
SDP
Catalog No. 5000348287
Encompass_Preferred
MRIA9 is a chemical probe and an ATP-competitive, pan Salt-Inducible kinase (SIK) and PAK2/3 inhibitor. It exhibits IC50 values of 516 nM, 180 nM, and 127 nM for SIK1, SIK2, and SIK3, respectively. This compound is intended for research use only.
  • Sensitizes SKOV3 cells to paclitaxel treatment by inducing pronounced apoptosis.
  • Significantly enhances cell death in HeLa cells when combined with paclitaxel.
  • Impedes centrosome function and causes mitotic spindle mispositioning in ovarian cancer cell lines.
  • Prevents centrosome disjunction during the late G2 phase.
  • Sensitizes ovarian cancer cells and patient-derived 3D-spheroids to paclitaxel treatment.
  • Shows high oral bioavailability (F = 75-80%).
Catalog No. 50-003-48287 Supplier Medchemexpress LLC Supplier No. HY13925350MG
MRIA9 is a chemical probe and an ATP-competitive, pan Salt-Inducible kinase (SIK) and PAK2/3 inhibitor. It exhibits IC50 values of 516 nM, 180 nM, and 127 nM for SIK1, SIK2, and SIK3, respectively. This compound is intended for research use only.
  • Sensitizes SKOV3 cells to paclitaxel treatment by inducing pronounced apoptosis.
  • Significantly enhances cell death in HeLa cells when combined with paclitaxel.
  • Impedes centrosome function and causes mitotic spindle mispositioning in ovarian cancer cell lines.
  • Prevents centrosome disjunction during the late G2 phase.
  • Sensitizes ovarian cancer cells and patient-derived 3D-spheroids to paclitaxel treatment.
  • Shows high oral bioavailability (F = 75-80%).