Learn More
Medchemexpress LLC HY-100370 5mg Medchemexpress, MRK-016 CAS:342652-67-9 Purity:>98%
SDP
Catalog No. 501931793
Encompass_Preferred
Medchemexpress, HY-100370 5mg MRK-016 CAS:342652-67-9 MRK-016 is a selective, orally bioavailable inverse agonist of GABA A α5 receptor , with an EC 50 of 3 nM for GABA A α5, and K i s of 0.83, 0.85, 0.77 and 1.4 nM for human GABA A  α1β3γ2, GABA A  α2β3γ2, GABA A  α3β3γ2, and GABA A  α5β3γ2, respectively; MRK-016 also readily penetrates the CNS. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-1793 Supplier Medchemexpress LLC Supplier No. HY1003705MG
Medchemexpress, HY-100370 5mg MRK-016 CAS:342652-67-9 MRK-016 is a selective, orally bioavailable inverse agonist of GABA A α5 receptor , with an EC 50 of 3 nM for GABA A α5, and K i s of 0.83, 0.85, 0.77 and 1.4 nM for human GABA A  α1β3γ2, GABA A  α2β3γ2, GABA A  α3β3γ2, and GABA A  α5β3γ2, respectively; MRK-016 also readily penetrates the CNS. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.