Learn More
Medchemexpress LLC HY-14174 5mg Medchemexpress, MRK-560 CAS:677772-84-8 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY141745MG
Encompass_Preferred
Medchemexpress, HY-14174 5mg MRK-560 CAS:677772-84-8 MRK-560 is a potent, orally bioavailable and brain-penetrant γ-secretase inhibitor [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2696