Learn More
Medchemexpress LLC MRK-623 | 461449-78-5 | 97.0% | 348.37 g·mol^-1 | C20H17FN4O | 50MG
SDP
Catalog No. 5000236387
Encompass_Preferred
MRK-623 is an orally active, high-affinity GABAA receptor agonist used for laboratory research. It shows anxiolytic effects and potent affinity for multiple GABAA subunits (reported Ki values: α1 0.85 nM; α2 3.7 nM; α3 4.0 nM; α5 0.53 nM).
Catalog No. 50-002-36387 Supplier Medchemexpress LLC Supplier No. HY12384050MG
MRK-623 is an orally active, high-affinity GABAA receptor agonist used for laboratory research. It shows anxiolytic effects and potent affinity for multiple GABAA subunits (reported Ki values: α1 0.85 nM; α2 3.7 nM; α3 4.0 nM; α5 0.53 nM).