Learn More
Medchemexpress LLC MRK-740 5mg | 2387510-80-5 | 5 MG
SDP
Catalog No. 5000227859
Encompass_Preferred
MRK-740 is a potent, selective, substrate-competitive inhibitor of the PRDM9 histone methyltransferase used as a research chemical probe. It shows biochemical potency (IC50 ≈ 80 nM) and reduces PRDM9-dependent H3K4 trimethylation in cells (~0.8 μM).
  • Potent PRDM9 inhibitor with IC50 ≈ 80 nM.
  • Reduces PRDM9-dependent H3K4 trimethylation in cellular assays (~0.8 μM).
  • High reported purity (99.84%).
  • Molecular weight 464.56; formula C25H32N6O3.
  • Supplied as a solid suitable for in vitro and cellular research.
Catalog No. 50-002-27859 Supplier Medchemexpress LLC Supplier No. HY1142095MG
MRK-740 is a potent, selective, substrate-competitive inhibitor of the PRDM9 histone methyltransferase used as a research chemical probe. It shows biochemical potency (IC50 ≈ 80 nM) and reduces PRDM9-dependent H3K4 trimethylation in cells (~0.8 μM).
  • Potent PRDM9 inhibitor with IC50 ≈ 80 nM.
  • Reduces PRDM9-dependent H3K4 trimethylation in cellular assays (~0.8 μM).
  • High reported purity (99.84%).
  • Molecular weight 464.56; formula C25H32N6O3.
  • Supplied as a solid suitable for in vitro and cellular research.