Learn More
Biomatik Corporation Mrka Polyclonal Antibody
Supplier:  Biomatik Corporation CAC11904
Encompass_Preferred
Mrka Polyclonal Antibody is a Klebsiella pneumoniae Polyclonal Antibody to mrkA. The Mrka Polyclonal Antibody has been validated for the following applications: ELISA. Host: Rabbit. Species: Klebsiella pneumoniae. Immunogen: Recombinant Klebsiella Pneumoniae Fimbrial Subunit Type 3 Protein (23-202Aa). Applications: ELISA. Target Synonyms: mrkA. Accession Number: P12267. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7537