Learn More
Medchemexpress LLC MRL-494 hydrochloride | 2699937-04-5 | 98.6% | 1 MG
SDP
Catalog No. 5000339355
Encompass_Preferred
MRL-494 hydrochloride is an antibacterial agent that functions as an inhibitor of β-barrel assembly machine A (BamA), remaining impervious to efflux and the outer membrane permeability barrier. It effectively inhibits both Gram-positive and Gram-negative bacteria by lethally disrupting the cytoplasmic membrane and inhibiting Outer Membrane Proteins (OMPs) biogenesis.
  • Antibacterial agent
  • Inhibitor of β-barrel assembly machine A (BamA)
  • Impervious to efflux and the outer membrane permeability barrier
  • Inhibits gram-positive bacteria
  • Inhibits gram-negative bacteria
  • Lethally disrupts the cytoplasmic membrane
  • Inhibits outer membrane proteins (OMPs) biogenesis
Catalog No. 50-003-39355 Supplier Medchemexpress LLC Supplier No. HY128773A1MG
MRL-494 hydrochloride is an antibacterial agent that functions as an inhibitor of β-barrel assembly machine A (BamA), remaining impervious to efflux and the outer membrane permeability barrier. It effectively inhibits both Gram-positive and Gram-negative bacteria by lethally disrupting the cytoplasmic membrane and inhibiting Outer Membrane Proteins (OMPs) biogenesis.
  • Antibacterial agent
  • Inhibitor of β-barrel assembly machine A (BamA)
  • Impervious to efflux and the outer membrane permeability barrier
  • Inhibits gram-positive bacteria
  • Inhibits gram-negative bacteria
  • Lethally disrupts the cytoplasmic membrane
  • Inhibits outer membrane proteins (OMPs) biogenesis