Learn More
Medchemexpress LLC MRL-494 hydrochloride | 2699937-04-5 | 98.6% | 1 ML
SDP
Catalog No. 5000339411
Encompass_Preferred
MRL-494 hydrochloride is an antibacterial agent that acts as an inhibitor of β-barrel assembly machine A (BamA). It is impervious to efflux and the outer membrane permeability barrier, demonstrating inhibitory effects on both Gram-positive and Gram-negative bacteria. MRL-494 lethally disrupts the cytoplasmic membrane and inhibits outer membrane proteins (OMPs) biogenesis by targeting BamA from outside the outer membrane.
  • Acts as an inhibitor of β-barrel assembly machine A (BamA).
  • Impervious to efflux and the outer membrane permeability barrier.
  • Effective against Gram-positive bacteria, including Staphylococcus aureus COL (MIC of 12.5 μM).
  • Effective against Gram-negative bacteria, including E. coli JCM158 (MIC of 25 μM).
  • Lethally disrupts the cytoplasmic membrane.
  • Inhibits outer membrane proteins (OMPs) biogenesis by targeting BamA.
Catalog No. 50-003-39411 Supplier Medchemexpress LLC Supplier No. HY128773A10MM/1ML
MRL-494 hydrochloride is an antibacterial agent that acts as an inhibitor of β-barrel assembly machine A (BamA). It is impervious to efflux and the outer membrane permeability barrier, demonstrating inhibitory effects on both Gram-positive and Gram-negative bacteria. MRL-494 lethally disrupts the cytoplasmic membrane and inhibits outer membrane proteins (OMPs) biogenesis by targeting BamA from outside the outer membrane.
  • Acts as an inhibitor of β-barrel assembly machine A (BamA).
  • Impervious to efflux and the outer membrane permeability barrier.
  • Effective against Gram-positive bacteria, including Staphylococcus aureus COL (MIC of 12.5 μM).
  • Effective against Gram-negative bacteria, including E. coli JCM158 (MIC of 25 μM).
  • Lethally disrupts the cytoplasmic membrane.
  • Inhibits outer membrane proteins (OMPs) biogenesis by targeting BamA.