Learn More
Medchemexpress LLC MRL-494 (hydrochloride) | 2699937-04-5 | 98.6% | 10 MG
SDP
Catalog No. 5000339374
Encompass_Preferred
MRL-494 hydrochloride is an antibacterial agent and an inhibitor of β-barrel assembly machine A (BamA). It is impervious to efflux and the outer membrane permeability barrier, making it effective against both Gram-positive and Gram-negative bacteria. This agent lethally disrupts the cytoplasmic membrane and inhibits outer membrane protein biogenesis by targeting BamA.
  • Inhibits β-barrel assembly machine A (BamA)
  • Effective against Gram-positive bacteria
  • Effective against Gram-negative bacteria
  • Disrupts cytoplasmic membrane
  • Inhibits outer membrane protein biogenesis
  • White to off-white solid
Catalog No. 50-003-39374 Supplier Medchemexpress LLC Supplier No. HY128773A10MG
MRL-494 hydrochloride is an antibacterial agent and an inhibitor of β-barrel assembly machine A (BamA). It is impervious to efflux and the outer membrane permeability barrier, making it effective against both Gram-positive and Gram-negative bacteria. This agent lethally disrupts the cytoplasmic membrane and inhibits outer membrane protein biogenesis by targeting BamA.
  • Inhibits β-barrel assembly machine A (BamA)
  • Effective against Gram-positive bacteria
  • Effective against Gram-negative bacteria
  • Disrupts cytoplasmic membrane
  • Inhibits outer membrane protein biogenesis
  • White to off-white solid