Learn More
Medchemexpress LLC HY-128773A 5mg Medchemexpress, MRL-494 (hydrochloride) CAS:0 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY128773A5MG
Encompass_Preferred
Medchemexpress, HY-128773A 5mg MRL-494 (hydrochloride) CAS:0 MRL-494 hydrochloride, an antibacterial agent, is a inhibitor of β-barrel assembly machine A (BamA) impervious to efflux and the outer membrane permeability barrier. MRL-494 hydrochloride can inhibits Gram-positive (MIC of 12.5 μM for Staphylococcus aureus COL) and Gram-negative (MIC of 25 μM for E. coli JCM158) bacterias. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-203-0128