Learn More
Aviva Systems Biology Corp MRM1 Antibody - C-terminal region (ARP41015_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP41015_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against MRM1. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the C terminal region of human MRM1. Alternate names: ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-57697