Learn More
VAZYME INTERNATIONAL LLC Vazyme mRNA Capture Beads, 2.0, 1 μm, Oligo(dT)-Coupled
Catalog No. NC3979705
Encompass
VAHTS mRNA Capture Beads 2.0 are 1 μm Oligo(dT)-coupled paramagnetic microspheres designed for isolating poly(A)+ RNA from purified total RNA. This magnetic separation technology allows for the isolation of intact mRNA from small sample volumes, removing the necessity for mRNA precipitation. The entire process can be finished within 1 hr.
  • Provides specific enrichment of mRNA, resulting in higher purity and reduced rRNA residue.
  • Works with various species (e.g., animals, plants, and fungi) and input sample amounts (from 10 ng to 12.5 μg).
Catalog No. NC3979705 Supplier VAZYME INTERNATIONAL LLC Supplier No. N40301
VAHTS mRNA Capture Beads 2.0 are 1 μm Oligo(dT)-coupled paramagnetic microspheres designed for isolating poly(A)+ RNA from purified total RNA. This magnetic separation technology allows for the isolation of intact mRNA from small sample volumes, removing the necessity for mRNA precipitation. The entire process can be finished within 1 hr.
  • Provides specific enrichment of mRNA, resulting in higher purity and reduced rRNA residue.
  • Works with various species (e.g., animals, plants, and fungi) and input sample amounts (from 10 ng to 12.5 μg).