Learn More
New England Biolabs, Inc. mRNA Decapping Enzyme – 2000 units
SDP
Catalog No. 502024040
Encompass
mRNA Decapping Enzyme catalyzes the removal of 7-methylguanosine cap (m7G) from 5 end of mRNA, producing 5 monophosphate and releasing m7GDP. mRNA Decapping Enzyme is capable of decapping mRNAs of various lengths and removes both Cap0 and Cap1 structures with similar efficiency. mRNA Decapping Enzyme also converts 5 triphosphate ends to 5 monophosphate, albeit with reduced efficiency. 5' monophosphorylated RNA can be exploited in a variety of downstream applications, including 5' RNA Ligase-mediated RACE, RNA-seq, and 5' -> 3' exonuclease digestion.
Catalog No. 50-202-4040 Supplier New England Biolabs, Inc. Supplier No. M0608S
mRNA Decapping Enzyme catalyzes the removal of 7-methylguanosine cap (m7G) from 5 end of mRNA, producing 5 monophosphate and releasing m7GDP. mRNA Decapping Enzyme is capable of decapping mRNAs of various lengths and removes both Cap0 and Cap1 structures with similar efficiency. mRNA Decapping Enzyme also converts 5 triphosphate ends to 5 monophosphate, albeit with reduced efficiency. 5' monophosphorylated RNA can be exploited in a variety of downstream applications, including 5' RNA Ligase-mediated RACE, RNA-seq, and 5' -> 3' exonuclease digestion.