Learn More
Takara Bio mRNA Interferase-MazF, 1000 units
SDP
Catalog No. NC1674865
Encompass
MazF is a toxin protein in the toxin-antitoxin module of E. coli. It possesses endoribonuclease activity and specifically cleaves single-stranded RNA at the 5' end of ACA sequences. This enzyme does not cleave double-stranded RNA, double-stranded DNA or single-stranded DNA.
Catalog No. NC1674865 Supplier Takara Bio Supplier No. 2415A
MazF is a toxin protein in the toxin-antitoxin module of E. coli. It possesses endoribonuclease activity and specifically cleaves single-stranded RNA at the 5' end of ACA sequences. This enzyme does not cleave double-stranded RNA, double-stranded DNA or single-stranded DNA.