Learn More
Apexbio Technology LLC MRNA MAG PURIF RNA LIB-96mg
SDP
Catalog No. 5000897391
Encompass_Preferred
mRNA Magnetic Purification kit for RNA library Prep is designed for applications in biomedical research focusing on mRNA isolation and purification Originating from cutting-edge molecular biology techniques this kit employs magnetic bead-based technology to selectively capture mRNA through oligo(dT) sequences that bind poly-A tails enabling high-purity isolation critical for downstream applications It is commonly utilized in in vitro activities for producing RNA libraries offering sensitivity in the nanomolar to low micromolar range This makes it an essential tool in various experimental settings including cell models animal studies and drug screening Experimental concentrations typically depend on the specific research design allowing flexibility in diverse research applications
Catalog No. 50-008-97391 Supplier Apexbio Technology LLC Supplier No. K181396
mRNA Magnetic Purification kit for RNA library Prep is designed for applications in biomedical research focusing on mRNA isolation and purification Originating from cutting-edge molecular biology techniques this kit employs magnetic bead-based technology to selectively capture mRNA through oligo(dT) sequences that bind poly-A tails enabling high-purity isolation critical for downstream applications It is commonly utilized in in vitro activities for producing RNA libraries offering sensitivity in the nanomolar to low micromolar range This makes it an essential tool in various experimental settings including cell models animal studies and drug screening Experimental concentrations typically depend on the specific research design allowing flexibility in diverse research applications