Learn More
Apexbio Technology LLC HyperScribe™ All in One mRNA Synthesis Kit (ARCA, T7, poly(A))(Synonyms: HyperScribe™ mRNA Kit, HyperScribe™ ARCA Kit, T7 ARCA-poly(A) mRNA Kit, All-in-One mRNA Synthesis Kit), 25 rxns.
SDP
Catalog No. 5000061402
Encompass_Preferred
HyperScribe All in One mRNA Synthesis Kit (ARCA T7 poly(A)) is designed for rapid in vitro generation of capped and polyadenylated mRNA transcripts Utilizing T7 RNA polymerase this system incorporates Anti-Reverse Cap Analog (ARCA) a Cap0 analog co-transcriptionally at the 5 terminus to confer mRNA stability and enhance translational efficiency After template DNA removal via DNase I treatment Poly(A) polymerase adds a poly(A) tail at the 3 -end further stabilizing the transcript and increasing translation initiation efficiency This kit is employed in research involving translation systems antisense RNA RNA interference RNA vaccines structural RNA studies and ribonuclease assays
Catalog No. 50-000-61402 Supplier Apexbio Technology LLC Supplier No. K106325
HyperScribe All in One mRNA Synthesis Kit (ARCA T7 poly(A)) is designed for rapid in vitro generation of capped and polyadenylated mRNA transcripts Utilizing T7 RNA polymerase this system incorporates Anti-Reverse Cap Analog (ARCA) a Cap0 analog co-transcriptionally at the 5 terminus to confer mRNA stability and enhance translational efficiency After template DNA removal via DNase I treatment Poly(A) polymerase adds a poly(A) tail at the 3 -end further stabilizing the transcript and increasing translation initiation efficiency This kit is employed in research involving translation systems antisense RNA RNA interference RNA vaccines structural RNA studies and ribonuclease assays