Learn More
System Biosciences LLC mRNAExpress™ GFP TranscriptmRNAExpress GFP transcript (positive control) = 10 µg
SDP
Catalog No. NC2008490
Encompass_Preferred
Deliver GFP mRNA to cells for visualization or as a positive control using this ready-to-transfect, pre-made mRNAExpress transcript. Save time when you want instant expression after transfection and use one of SBI’s pre-made, ready-to-transfect mRNAs. Like all of our pre-made mRNAs, the mRNAExpress™ GFP Transcript contains a number of different features to enhance performance.
Catalog No. NC2008490 Supplier System Biosciences LLC Supplier No. MR700A1
Deliver GFP mRNA to cells for visualization or as a positive control using this ready-to-transfect, pre-made mRNAExpress transcript. Save time when you want instant expression after transfection and use one of SBI’s pre-made, ready-to-transfect mRNAs. Like all of our pre-made mRNAs, the mRNAExpress™ GFP Transcript contains a number of different features to enhance performance.