Learn More
Aviva Systems Biology Corp MRO Antibody - C-terminal region (ARP60588_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP60588_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against MRO. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the C terminal region of human MRO. Alternate names: B29, C18orf3, FLJ30140ApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-287-095