Learn More
enQuire Bioreagents Calponin-1 Anti-Human Monoclonal [Clone SPM169], Size: 100ug
Supplier:  enQuire Bioreagents 4363MSM1P1
Encompass_Preferred
Product Type: Antibody. Application: IHC. Conjugate/Tag: Unconjugated. Clone: MRP1/1343. Clonality: Monoclonal. Host: Mouse. Isotype: IgG1. Positive Control: A549 cells. Colon, Endometrium, Ovary. Testis or Tonsil. Cellular Staining: Cytoplasmic and Cell Surface. Buffer: 10mM PBS with 0.05% BSA and 0.05% Azide. Concentration: 200 ug/ml. Purity: Protein A/G Purified. Immunogen: Recombinant human full-length ABCC1. Storage: Short term: Store at 2-4C. Long term, aliquot and store at -20C. MW: 190kDa.Chromosome: 16p13.11. Gene Symbol: ABCC1. Gene ID: 4363. Ensemble: ENSG00000278183. Uniprot: XP_016878731.
Catalog No. 50-188-4943