Learn More
Enzo Life Sciences MRP1 (human) monoclonal antibody (MRPm6) (250µg)
SDP
Supplier:  Enzo Life Sciences ALX801013C250
Encompass_Preferred
Clone: MRPm6. Isotype: IgG1. Immunogen: Recombinant human MRP1 (multidrug resistance-associated protein 1) (aa 1294-1531). Species reactivity: Human. Specificity: Recognizes the internal epitope (P1511SDLLQQRGL1520) of human MRP1. Applications: Flow Cytometry, ICC, IHC (FS), WB. Formulation: Liquid. In serum free culture supernatant containing 0.7% BSA and 0.1% sodium azide.
Catalog No. 50-200-9689