Learn More
Enzo Life Sciences MRP1 (human) monoclonal antibody (QCRL-1) (1ml)
SDP
Supplier:  Enzo Life Sciences ALX801010L001
Encompass_Preferred
Clone: QCRL-1. Isotype: IgG1. Immunogen: Membranes prepared from the human small cell lung cancer cell line H69AR. Species reactivity: Human. Specificity: Recognizes the linear internal epitope (S918SYSGDI924) of human MRP1 (multidrug resistance-associated protein 1), overexpressed in various human non-P-glycoprotein MDR tumor cell lines. Applications: Flow Cytometry, ICC, IHC (PS), IP, WB. Formulation: Liquid. 0.22μm-filtered solution in PBS containing 0.7% BSA and 0.1% sodium azide.
Catalog No. 50-200-9686