Learn More
Enzo Life Sciences MRP1 monoclonal antibody (MRPr1) (250µg)
SDP
Supplier:  Enzo Life Sciences ALX801007C250
Encompass_Preferred
Clone: MRPr1. Isotype: IgG2a. Immunogen: Recombinant human MRP1 (multidrug resistance-associated protein 1) (aa 192-360). Species reactivity: Human, Mouse. Specificity: Recognizes the internal epitope (G238SDLWSLNKE247) of MRP1, overexpressed in various human non-P-glycoprotein MDR tumor cell lines. Applications: Flow Cytometry, ICC, IHC (FS), IHC (PS), WB. Formulation: Liquid. In serum free culture supernatant containing 0.7% BSA and 0.1% sodium azide.
Catalog No. 50-200-9685