Learn More
Enzo Life Sciences MRP2 (human) monoclonal antibody (M2I-4) (250µg)
SDP
Supplier:  Enzo Life Sciences ALX801015C250
Encompass_Preferred
Clone: M2I-4. Isotype: IgG1. Immunogen: Recombinant human MRP2 (multidrug resistance-associated protein 2; cMOAT) (aa 215-310). Species reactivity: Human. Specificity: Recognizes an internal epitope of human MRP2. Applications: ICC, IHC (FS), WB. Formulation: Liquid. In serum free culture supernatant containing 0.7% BSA and 0.1% sodium azide.
Catalog No. 50-200-9691