Learn More
Enzo Life Sciences MRP2 monoclonal antibody (M2III-5) (125µg)
SDP
Supplier:  Enzo Life Sciences ALX801037C125
Encompass_Preferred
Clone: M2III-5. Isotype: IgG2b. Immunogen: Recombinant C-terminal rat MRP2 (multidrug resistance-associated protein 2; cMOAT) (aa 1339-1541). Species reactivity: Human, Mouse, Rat. Specificity: Recognizes an internal epitope of MRP2. Applications: ICC, IHC (FS), IHC (PS), WB. Formulation: Liquid. In serum free culture supernatant containing 0.7% BSA and 0.1% sodium azide.
Catalog No. 50-200-9706