Learn More
Aviva Systems Biology Corp MRPL1 Antibody - N-terminal region (ARP62868_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP62868_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against MRPL1. It was validated on Western Blot. . Alternate names: BM022, FLJ96680, L1MT, MRP-L1ApplicationWB. Species reactivity: Dog, Guinea Pig, Horse, Human, Rabbit, Rat
Catalog No. 50-287-793