Learn More
Aviva Systems Biology Corp MRPL10 Antibody - N-terminal region (ARP52830_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP52830_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against MRPL10. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human MRPL10. Alternate names: MGC17973, MRP-L8, RPML8, L10MT, MRPL8, MRP-L10ApplicationWB. Species reactivity: Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-60843