Learn More
Aviva Systems Biology Corp MRPL13 Antibody - middle region (ARP54978_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP54978_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against MRPL13. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human MRPL13. Alternate names: L13, L13mt, RPL13, RPML13, L13AApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Zebrafish
Catalog No. 50-014-53903