Learn More
Aviva Systems Biology Corp MRPL15 Antibody - C-terminal region (ARP46192_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP46192_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against MRPL15. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human MRPL15. Alternate names: HSPC145, MRP-L7, RPML7, L15mt, MRP-L15ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Zebrafish
Catalog No. 50-014-59265