Learn More
Aviva Systems Biology Corp MRPL24 Antibody - N-terminal region (ARP41010_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP41010_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against MRPL24. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human MRPL24. Alternate names: FLJ20917, MGC22737, MGC9831, MRP-L18, L24mt, MRP-L24ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-53257