Learn More
Aviva Systems Biology Corp MRPL3 Antibody - N-terminal region (ARP62688_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP62688_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against MRPL3. It was validated on Western Blot. . Alternate names: MRL3, RPML3ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-287-741