Learn More
Aviva Systems Biology Corp MRPL37 Antibody - N-terminal region (ARP56924_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP56924_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against MRPL37. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human MRPL37. Alternate names: MGC878, MRP-L2, MRPL2, RPML2, L37mt, MRP-L37ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-62036