Learn More
Aviva Systems Biology Corp MRPL38 Antibody - C-terminal region (ARP62474_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP62474_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against MRPL38. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the C-terminal region of Human MRPL38. Alternate names: FLJ13996, HSPC262, KIAA1863, MGC4810, MRP-L3, RPML3, L38MT, MRP-L38ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-287-692