Learn More
Aviva Systems Biology Corp MRPL43 Antibody - N-terminal region (ARP62689_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP62689_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against MRPL43. It was validated on Western Blot. . Alternate names: MGC17989, MGC48892, bMRP36aApplicationWB. Species reactivity: Cow, Guinea Pig, Horse, Human, Mouse, Rat
Catalog No. 50-287-742