Learn More
Aviva Systems Biology Corp MRPL45 Antibody - C-terminal region (ARP60477_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP60477_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against MRPL45. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the C terminal region of human MRPL45. Alternate names: MGC11321, L45mt, MRP-L45ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-287-057