Learn More
Aviva Systems Biology Corp MRPL49 Antibody - N-terminal region (ARP60087_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP60087_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against MRPL49. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the N terminal region of human MRPL49. Alternate names: C11orf4, L49mt, MGC10656, NOF, NOF1, MRP-L49ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-286-936