Learn More
Aviva Systems Biology Corp Mrpl55 Antibody - N-terminal region (ARP53388_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP53388_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Mrpl55. It was validated on Western Blot. . Alternate names: 2810038N09RikApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-285-832