Learn More
Aviva Systems Biology Corp MRPL55 Antibody - middle region (ARP53389_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP53389_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against MRPL55. It was validated on Western Blot. . Alternate names: AAVG5835, DKFZp686D1387, L55nt, MGC61802, MRP-L55, PRO19675ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Yeast
Catalog No. 50-285-833