Learn More
Aviva Systems Biology Corp MRPS12 Antibody - N-terminal region (ARP46301_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP46301_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against MRPS12. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human MRPS12. Alternate names: MPR-S12, MT-RPS12, RPMS12, RPS12, RPSM12ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Rat
Catalog No. 50-014-53469