Learn More
Aviva Systems Biology Corp MRPS15 Antibody - C-terminal region (ARP39660_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP39660_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against MRPS15. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human MRPS15. Alternate names: DC37, FLJ11564, MPR-S15, RPMS15, S15mtApplicationWB. Species reactivity: Human
Catalog No. 50-014-53148