Learn More
Aviva Systems Biology Corp MRPS15 Antibody - N-terminal region (ARP33299_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP33299_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against MRPS15. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the N terminal region of human MRPS15. Alternate names: DC37, FLJ11564, MPR-S15, RPMS15, S15mtApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-52899